Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: This MR54 monoclonal antibody reacts with human Trop-2, which is also known as Tumor-Associated Calcium Signal Transducer 2 (TACSTD2) and EGP-1. This type I transmembrane glycoprotein is expressed on trophoblast (syncytiotrophoblast and cytotrophoblast), epithelial, and most human carcinoma cells. Trop-2 plays a role in calcium-mediated signaling. Trop-2 has also been reported as an oncogene, with expression being correlated with tumorigenesis, metastasis, and anchorage-independent growth.
Applications Reported: This MR54 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of formalin-fixed paraffin embedded (FFPE) tissue sections (IHC-P), and immunocytochemistry (ICC).
Applications Tested: This MR54 antibody has been tested by flow cytometry and immunocytochemistry on MCF7 cells. This can be used at less than of equal to 10 µg/mL. For immunohistochemistry of formalin-fixed paraffin embedded tissue, either low or high pH antigen retrieval can be used, although high pH antigen retrieval results in better staining intensity. Is is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: 40kD glycoprotein, identified by monoclonal antibody GA733; Cell surface glycoprotein Trop-2; cell surface glycoprotein TROP2; epithelial glycoprotein-1; GA733; gastrointestinal tumor-associated antigen GA7331; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; Pancreatic carcinoma marker protein GA733-1; pancreatic carcinoma marker protein GA7331; TACD2; truncated TACSTD2; Tumor-associated calcium signal transducer 2
Gene Aliases: EGP-1; EGP1; GA733-1; GA7331; GP50; M1S1; TACSTD2; TROP2
UniProt ID: (Human) P09758
Entrez Gene ID: (Human) 4070
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support