Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPNSEPASL LELFNSIATQ GELVRSLKAG NASKDEIDSA VKMLVSLKMS YKAAAGEDYK ADCPPGNPAP TSNHGPDATE AEEDFVDPWT VQTSSAKGID YDKLIVRFGS SKIDKELINR IERATGQRPH HFLRRGIFFS HRDMNQVLDA YENKKPFYLY TGRGPSSEAM HVGHLIPFIF TKWLQDVFNV PLVIQMTDDE KYLWKDLTLD QAYSYAVENA KDIIACGFDI NKTFIFSDLD YMGMSSGFYK NVVKIQKHVT FNQVKGIFGF TDSDCIGKIS FPAIQAAPSF SNSFPQIFRD RTDIQCLIPC AIDQDPYFRM TRDVAPRIGY PKPALLHSTF FPALQGAQTK MSASDPNSSI FLTDTAKQI (1-368 aa encoded by BC017489 )
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. Tryptophanyl-tRNA synthetase (WARS) catalyzes the aminoacylation of tRNA(trp) with tryptophan and is induced by interferon. Tryptophanyl-tRNA synthetase belongs to the class I tRNA synthetase family. Four transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hWRS; IFI 53; IFP 53; IFP53; Interferon-induced protein 53; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; Tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, cytoplasmic; WARS protein
Gene Aliases: GAMMA-2; IFI53; IFP53; TrpRS; WARS; WARS1; WRS
UniProt ID: (Human) P23381, (Mouse) P32921
Entrez Gene ID: (Human) 7453, (Rat) 314442, (Mouse) 22375
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support