Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
FIGURE: 1 / 3
Tryptophan hydroxylase (TPH) catalyzes the 5-hydroxylation of tryptophan, which is the first step in the biosynthesis of indoleamines (serotonin and melatonin) (Martinez et al., 2001). In mammals, serotonin biosynthesis occurs predominantly in neurons which originate in the Raphe nuclei of the brain, and melatonin synthesis takes place within the pineal gland. Although TPH catalyzes the same reaction within the Raphe nuclei and the pineal gland, TPH activity is rate-limiting for serotonin but not melatonin biosynthesis. Serotonin functions mainly as a neurotransmitter, whereas melatonin is the principal hormone secreted by the pineal gland. The activity of TPH is enhanced by phosphorylation by cAMP-dependent protein kinase (PKA) and Ca2+/calmodulin kinase II (CAM K II) (Jiang et al., 2000; Johansen et al., 1996). Both PKA and CAM K II phosphorylate Ser58 which lies within the regulatory domain of TPH (Kuhn et al., 1997).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; MGC119994; Neuronal tryptophan hydroxylase; Tryptophan 5-hydroxylase 1; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 1; Tryptophan 5-monooxygenase 2; tryptophan hydroxylase (tryptophan 5-monooxygenase)
Gene Aliases: ADHD7; AU043594; NTPH; TPH; TPH1; TPH2; TPRH; TRPH
UniProt ID: (Human) P17752, (Human) Q8IWU9, (Mouse) P17532, (Rat) P09810, (Mouse) Q8CGV2, (Rat) Q8CGU9
Entrez Gene ID: (Human) 7166, (Human) 121278, (Mouse) 21990, (Rat) 24848, (Mouse) 216343, (Rat) 317675
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support