Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Control: Raji, THP-1, HL-60, TWIST1 transfected 293T lysates, human TWIST1-transfected 293T, Jurkat, OECM1, CL1-5, Toledo, SU-DHL-5, HT, DB, RL95-2, MDA-MB-231
Predicted Reactivity: Japanese Medaka (95%), Xenopus laevis (97%), Chicken (100%), Chimpanzee (100%), Bovine (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Twist1/Twist2 encodes a basic helix-loop-helix (bHLH) transcription factor that plays an important role in embryonic development. The encoded protein forms both homodimers and heterodimers that bind to DNA E box sequences and regulate the transcription of genes involved in cranial suture closure during skull development. This protein may also regulate neural tube closure, limb development and brown fat metabolism. Twist1/Twist2 is hypermethylated and overexpressed in multiple human cancers, and the encoded protein promotes tumor cell invasion and metastasis. Mutations in this gene cause Saethre-Chotzen syndrome in human patients, which is characterized by craniosynostosis, ptosis and hypertelorism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-HLH DNA binding protein; bHLHa38; bHLHa39; charlie chaplin; Class A basic helix-loop-helix protein 38; Class A basic helix-loop-helix protein 39; dermis expressed 1; Dermis-expressed protein 1; Dermo-1; dermo-1 protein; H-twist; M-twist; MGC117334; Pluridigite; polydactyly EMS; Skam10Jus; twist basic helix-loop-helix transcription factor 1; twist basic helix-loop-helix transcription factor 2; twist gene homolog 1; twist homolog 1; twist homolog 2; TWIST homolog of drosophila; twist-related bHLH protein Dermo1; Twist-related protein 1; Twist-related protein 2
Gene Aliases:
AA960487; ACS3; AMS; BBRSAY; BHLHA38; BHLHA39; BPES2; BPES3; CRS; CRS1; CSO; DERMO1; FFDD3; M-Twist; Pde; pdt; SCS; SETLSS; Ska10; Ska
UniProt ID: (Human) Q15672, (Human) Q8WVJ9, (Mouse) P26687, (Mouse) Q9D030, (Rat) P97831
Entrez Gene ID: (Human) 7291, (Human) 117581, (Mouse) 22160, (Rat) 85489, (Mouse) 13345, (Rat) 59327
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support