Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: FPRACVSSKN LMEKECCPPW SGDRSPCGQL SGRGSCQNIL LSNAPLGPQF PFTGVDDRES WPSVFYNRTC QCSGNFMGFN CGNCKFGFWG PNCTERRLLV RRNIFDLSAP EKDKFFAYLT LAKHTISSDY VIPIGTYGQM KNGSTPMFND INIYDLFVWM HYYVSMDALL GGSEIWRDID FAHEAPAFLP WHRLFLLRWE QEIQKLTGDE NFTIPYWDWR DAEKCDICTD EYMGGQHPTN PNLLSPASFF SSWQIVCSRL EEYNSHQSLC NGTPEGPLRR NPGNHDKSRT PRLPSSADVE FCLSLTQYES GSMDKAANFS F; Positive Samples: Rat skin; Cellular Location: Melanosome membrane, Single-pass type I membrane protein
The enzyme encoded by this gene catalyzes the first 2 steps, and at least 1 subsequent step, in the conversion of tyrosine to melanin. The enzyme has both tyrosine hydroxylase and dopa oxidase catalytic activities, and requires copper for function. Mutations in this gene result in oculocutaneous albinism, and nonpathologic polymorphisms result in skin pigmentation variation. The human genome contains a pseudogene similar to the 3' half of this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LB24-AB; Monophenol monooxygenase; oculocutaneous albinism IA; SK29-AB; Tumor rejection antigen AB; Tyrosinase
Gene Aliases: ATN; C; CMM8; OCA1; OCA1A; OCAIA; SHEP3; TYR
UniProt ID: (Human) P14679
Entrez Gene ID: (Human) 7299, (Rat) 308800
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support