Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ACRHGDRCS RLHNKPTFSQ TIALLNIYRN PQNSSQSADG LRCAVSDVEM QEHYDEFFEE VFTEMEEKYG EVEEMNVCDN LGDHLVGNVY VKFRREEDAE KAVIDLNNRW FNGQPIHAEL SPVTDFREAC CRQYEMGECT RGGFCNFMHL KPISRELRRE LYGRRRKKHR SRSRSRERRS RSRDRGRGGG GGGGGGGGGR ERDRRRSRDR ERSGRF (26-240 aa encoded by BC001923)
This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Splicing factor U2AF 35 kDa subunit; splicing factor U2AF 35kDa subunit; U2 auxiliary factor 35 kDa subunit; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF),35 kDa; U2 small nuclear ribonucleoprotein auxillary factor, 35-KD subunit; U2 small nuclear RNA auxiliary factor 1; U2 small nuclear RNA auxillary factor 1; U2 snRNP auxiliary factor small subunit; U2(RNU2) small nuclear RNA auxiliary factor binding protein
Gene Aliases: 2010107D16Rik; 35kDa; FP793; RN; RNU2AF1; U2AF1; U2AF35; U2AFBP
UniProt ID: (Human) Q01081, (Mouse) Q9D883
Entrez Gene ID: (Human) 7307, (Mouse) 108121, (Rat) 687575
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support