Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: RGMPQEERLS RYQQFKDFQR RILVATNLFG RGMDIERVNI AFNYDMPEDS DTYLHRVARA GRFGTKGLAI TFVSDENDAK ILNDVQDRFE VNISELPDEI DISSYIEQTR; Positive Samples: 293T, HeLa, LO2, SKOV3, K-562, SW620; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle
This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 56 kDa U2AF65-associated protein; ATP-dependent RNA helicase p47; D17H6S81E-1; D20H6S81e; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B; DEAD box protein UAP56; HLA-B associated transcript 1; HLA-B associated transcript 1A; HLA-B-associated transcript 1 protein; HLA-B-associated transcript 1A; Nuclear RNA helicase; nuclear RNA helicase (DEAD family); nuclear RNA helicase Bat1; spliceosome RNA helicase BAT1; Spliceosome RNA helicase DDX39B
Gene Aliases: 0610030D10Rik; AI428441; Bat-1; BAT1; Bat1a; D17H6S81E; D17H6S81E-1; D6S81E; D6S81Eh; DDX39B; p47; UAP56
UniProt ID: (Human) Q13838, (Rat) Q63413, (Mouse) Q9Z1N5
Entrez Gene ID: (Human) 7919, (Rat) 114612, (Mouse) 53817
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support