Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anthracycline-associated resistance ARX; FLJ13058; SUMO-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; SUMO1 activating enzyme subunit 2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin-like 1 (sentrin) activating enzyme E1B; ubiquitin-like 1 (sentrin) activating enzyme subunit 2; Ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; Ubiquitin-like modifier-activating enzyme 2
Gene Aliases: AA986091; ARX; HRIHFB2115; SAE2; UBA1; UBA2; Ubl1a2; UBLE1B
UniProt ID: (Human) Q9UBT2, (Mouse) Q9Z1F9
Entrez Gene ID: (Human) 10054, (Rat) 308508, (Mouse) 50995
Molecular Function:
DNA-binding transcription factor
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support