Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase subunit KPC2; Glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; putative glialblastoma cell differentiation-related; putative glialblastoma cell differentiation-related protein; RP11-432J22.3; testicular secretory protein Li 64; UBA domain-containing protein 1; ubiquitin associated domain containing 1; Ubiquitin-associated domain-containing protein 1
Gene Aliases: 1110033G07Rik; AA407978; GBDR1; Gdbr1; KPC2; UBAC1; UBADC1
UniProt ID: (Human) Q9BSL1, (Mouse) Q8VDI7
Entrez Gene ID: (Human) 10422, (Mouse) 98766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support