Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DGVLERRPYE DQGLGETTPL TIICQPMQPL RVNSQPGPQK RCLFVCRHGE RMDVVFGKYW LSQCFDAKGR YIRTNLNMPH SLPQRSGGFR DYEKDAPI
Sts1 encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cbl-interacting protein p70; Cbl-interacting protein Sts-1; MGC15437; nm23-phosphorylated unknown substrate; SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling 1, nm23-phosphorylated unknown substrate; STS-1; Suppressor of T-cell receptor signaling 1; T-cell ubiquitin ligand 2; TULA-2; Tyrosine-protein phosphatase STS1/TULA2; Ubiquitin-associated and SH3 domain-containing protein B
Gene Aliases: KIAA1959; p70; STS-1; STS1; TULA-2; TULA2; UBASH3B
UniProt ID: (Human) Q8TF42
Entrez Gene ID: (Human) 84959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support