Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LHLSFLQNAA KLYATVYCIP FAEEDLSADA LLNILSEVKI QEFKPSNKVV QTDETARKPD HVPISSEDER NAIFQLEKAI LSNEAT
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 87%.
UBA6 (UBE1L2, MOP-4) is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E1-L2; Monocyte protein 4; MOP-4; UBA6, ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme 6; ubiquitin-activating enzyme E1-like 2; Ubiquitin-activating enzyme E1-like protein 2; ubiquitin-like modifier activating enzyme 6; Ubiquitin-like modifier-activating enzyme 6
Gene Aliases: E1-L2; MOP-4; MOP4; UBA6; UBE1L2
UniProt ID: (Human) A0AVT1
Entrez Gene ID: (Human) 55236
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support