Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTPARRRL MRDFKRLQED PPVGVSGAPS ENNIMQWNAV IFGPEGTPFE DGTFKLVIEF SEEYPNKPPT VRFLSKMFHP NVYADGSICL DILQNRWSPT YDVSSILTSI QSLLDEPNPN SPANSQAAQL YQENKREYEK RVSAIVEQSW NDS (1-152 aa encoded by BC005979)
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; HR6B; RAD6 homolog B; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; Ubiquitin-protein ligase B
Gene Aliases: 2610301N02Rik; E2-14k; E2-17kDa; HHR6B; HR6B; mHR6B; RAD6B; UBC2; UBCH1; UBE2B
UniProt ID: (Human) P63146, (Rat) P63149, (Mouse) P63147
Entrez Gene ID: (Human) 7320, (Rat) 81816, (Mouse) 22210
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support