Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro able to promote polyubiquitination using all 7 ubiquitin Lys residues, but may prefer 'Lys-11' and 'Lys-48'-linked polyubiquitination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme D4; HBUCE1; Ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2D 4 (putative); Ubiquitin-conjugating enzyme E2 D4; ubiquitin-conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme HBUCE1; Ubiquitin-protein ligase D4
Gene Aliases: HBUCE1; UBCH5D; UBE2D4
UniProt ID: (Human) Q9Y2X8
Entrez Gene ID: (Human) 51619
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support