Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTEAQRVDD SPSTSGGSSD GDQRESVQQE PEREQVQPKK KEGKISSKTA AKLSTSAKRI QKELAEITLD PPPNCSAGPK GDNIYEWRST ILGPPGSVYE GGVFFLDITF SPDYPFKPPK VTFRTRIYHC NINSQGVICL DILKDNWSPA LTISKVLLSI CSLLTDCNPA DPLVGSIATQ YMTNRAEHDR MARQWTKRYA T
ubiquitin-conjugating enzyme E2 (UBE2E2) is a ubiquitin conjugating enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme E2; FLJ25157; UbcH8; Ubiquitin carrier protein E2; ubiquitin conjugating enzyme E2E 2; Ubiquitin-conjugating enzyme E2 E2; ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast); Ubiquitin-protein ligase E2
Gene Aliases: UBCH8; UBE2E2
UniProt ID: (Human) Q96LR5
Entrez Gene ID: (Human) 7325
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support