Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSSPSPGKRR MDTDVVKLIE SKHEVTILGG LNEFVVKFYG PQGTPYEGGV WKVRVDLPDK YPFKSPSIGF MNKIFHPNID EASGTVCLDV INQTWTALYD LTNIFESFLP QLLAYPNPID PLNGDAAAMY LHRPEEYKQK IKEYIQKYAT EEALKEQEEG TGDSSSESSM SDFSEDEAQD MEL
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein sequence is 100% identical to the mouse homolog and 98% identical to the frog and zebrafish homologs. Three alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme H; E2 ubiquitin-conjugating enzyme H; GID complex subunit 3, UBC8 homolog; UbcH2; Ubiquitin carrier protein H; ubiquitin conjugating enzyme E2H; Ubiquitin-conjugating enzyme E2 H; Ubiquitin-conjugating enzyme E2-20K; ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8); ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast); Ubiquitin-protein ligase H
Gene Aliases: E2-20K; GID3; UBC8; UBCH; UBCH2; UBE2H
UniProt ID: (Human) P62256
Entrez Gene ID: (Human) 7328
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support