Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MMASMRVVKE LEDLQKKPPP YLRNLSSDDA NVLVWHALLL PDQPPYHLKA FNLRISFPPE YPFKPPMIKF TTKIYHPNVD ENGQICLPII SSENWKPCTK TCQVLEALNV LVNRPNIREP LRMDLADLLT QNPELFRKNA EEFTLRFGVD RPS
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to function in the E6/E6-ap-induced ubiquitination of p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme L6; MGC40331; retinoic acid induced gene B protein; Retinoic acid-induced gene B protein; RIG-B; UbcH8; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2L 6; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6; Ubiquitin/ISG15-conjugating enzyme E2 L6
Gene Aliases: RIG-B; UBCH8; UBE2L6
UniProt ID: (Human) O14933
Entrez Gene ID: (Human) 9246
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support