Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEDD8 carrier protein; NEDD8 protein ligase; NEDD8-conjugating enzyme Ubc12; UBC12 homolog, yeast; ubiquitin C, related sequence 2; ubiquitin carrier protein M; ubiquitin conjugating enzyme E2M; Ubiquitin-conjugating enzyme E2 M; ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast); ubiquitin-protein ligase M; yeast UBC12 homolog
Gene Aliases: 2510040H03Rik; hUbc12; UBC-RS2; UBC12; UBE2M
UniProt ID: (Human) P61081, (Mouse) P61082
Entrez Gene ID: (Human) 9040, (Dog) 484221, (Mouse) 22192
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support