Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNSNVENLP PHIIRLVYKE VTTLTADPPD GIKVFPNEED LTDLQVTIEG PEGTPYAGGL FRMKLLLGKD FPASPPKGYF LTKIFHPNVG ANGEICVNVL KRDWTAELGI RHVLLTIKCL LIHPNPESAL NEEAGRLLLE NYEEYAARAR LLTEIHGGAG GPSGRAEAGR ALASGTEASS TDPGAPGGPG GAEGPMAKKH AGERDKKLAA KKKTDKKRAL RRL (1-222 aa encoded by BC004236)
This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme S; E2-EPF; E2-EPF5; Ubiquitin carrier protein S; ubiquitin conjugating enzyme E2S; Ubiquitin-conjugating enzyme E2 S; ubiquitin-conjugating enzyme E2-24 kD; Ubiquitin-conjugating enzyme E2-24 kDa; Ubiquitin-conjugating enzyme E2-EPF5; ubiquitin-conjugating enzyme E2S; Ubiquitin-protein ligase S
Gene Aliases: 0910001J09Rik; 6720465F12Rik; AA409170; E2-EPF; E2EPF; EPF5; OK/SW-cl.73; RGD1564746; UBE2S
UniProt ID: (Human) Q16763, (Rat) B5DFI8, (Mouse) Q921J4
Entrez Gene ID: (Human) 27338, (Rat) 292588, (Mouse) 77891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support