Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
UBE2T is involved in the covalent conjugation of ubiquitin to proteins regulates diverse cellular pathways and proteins. Ubiquitin is transferred to a target protein through a concerted action of a ubiquitin-activating enzyme (E1), a ubiquitin-conjugating enzyme (E2), such as UBE2T, and a ubiquitin ligase (E3). UBE2T catalyzes the covalent attachment of ubiquitin to protein substrates. Functionally, UBE2T accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins, catalyzes monoubiquitination, is involved in mitomycin-C (MMC)-induced DNA repair, and catalyzES monoubiquitination of FANCD2, a key step in the DNA damage pathway. Defects in the UBE2T gene have been associated with Fanconi anemia of complementation group T.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 50 protein; E2 ubiquitin-conjugating enzyme T; HSPC150 protein similar to ubiquitin-conjugating enzyme; Ubiquitin carrier protein T; ubiquitin conjugating enzyme E2T; Ubiquitin-conjugating enzyme E2 T; ubiquitin-conjugating enzyme E2T (putative); Ubiquitin-protein ligase T
Gene Aliases: FANCT; HSPC150; PIG50; UBE2T
UniProt ID: (Human) Q9NPD8
Entrez Gene ID: (Human) 29089
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support