Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAATTGSGVK VPRNFRLLEE LEEGQKGVGD GTVSWGLEDD EDMTLTRWTG MIIGPPRTIY ENRIYSLKIE CGPKYPEAPP FVRFVTKINM NGVNSSNGVV DPRAISVLAK WQNSYSIKVV LQELRRLMMS KENMKLPQPP EGQCYSN
The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CROC-1; CROC-1UBE2V; CROC1B; DJ1185N5.1.3; DNA-binding protein; KUA-UEV; TRAF6-regulated IKK activator 1 beta Uev1A; ubiquitin conjugating enzyme E2 variant 1; Ubiquitin-conjugating enzyme E2 variant 1; UEV-1; UEV-1CIR1
Gene Aliases: CIR1; CROC-1; CROC1; P/OKcl.19; UBE2V; UBE2V1; UEV-1; UEV1; UEV1A
UniProt ID: (Human) Q13404
Entrez Gene ID: (Human) 7335
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support