Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLVIESLRNS KYIKKFPLLE NTFKADSSSA WSAVKVLYQP CIKSRNEKLV SLWESDISVH PLTLPSATCL ELLL
Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase E3D; HECT-type E3 ubiquitin transferase E3D; UBCH10 binding protein with a hect-like domain; UbcH10-binding protein with a HECT-like domain; ubiquitin-conjugating enzyme E2C binding protein; Ubiquitin-conjugating enzyme E2C-binding protein
Gene Aliases: C6orf157; H10BH; UBE2CBP; UBE3D; YJR141W
UniProt ID: (Human) Q7Z6J8
Entrez Gene ID: (Human) 90025
Molecular Function:
ubiquitin-protein ligase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support