Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLAEALIKV FVDIEFTGDP HQFEQKFNYR RPMYPILRYM WGTDTYRESI KDLADYASKN LEAMNPPLFL RFLNLLMNDA IFLLDEAIQY LSKIKIQQIE KDRGEWDSLT PEARREKEAG LQMFGQLARF HNIMSNETIG TLAFLTSEIK SLFVHPFLAE RIISMLNYFL QHLVGPKMGA LKVKDFSEFD FKPQQLVSDI CTIYLNLGDE ENFCATVPKD GRSYSPTLFA QTVRVLKKIN KPGNMIMAFS NLAERIKSLA DLQQQEEETY ADACDEFLDP IMSTLMCDPV VLPSSRVTVD RSTIARHLLS DQTDPFNRSP LTMDQIRPNT ELKEKIQRWL AERKQQKEQL E (717-1066 aa encoded by BC112367)
Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING-type E3 ubiquitin transferase E4 A; Ubiquitin conjugation factor E4 A; ubiquitination factor E4A (UFD2 homolog, yeast); ubiquitination factor E4A, UFD2 homolog
Gene Aliases: 4732444G18Rik; 9930123J21Rik; E4; KIAA0126; UBE4A; UBOX2; UFD2; UFD2b
UniProt ID: (Human) Q14139, (Mouse) E9Q735
Entrez Gene ID: (Human) 9354, (Mouse) 140630
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support