Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: HUVEC, Mouse brain, Mouse kidney, Rat liver, Rat brain. The target is usually found in the following locations: Cytoplasm.
Immunogen sequence: MTDQENNNNI SSNPFAALFG SLADAKQFAA IQKEQLKQQS DELPASPDDS DNSVSESLDE FDYSVAEISR SFRSQQEICE QLNINHMIQR IFLITLDNSD
Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING-type E3 ubiquitin transferase E4 A; Ubiquitin conjugation factor E4 A; ubiquitination factor E4A (UFD2 homolog, yeast); ubiquitination factor E4A, UFD2 homolog
Gene Aliases: 4732444G18Rik; 9930123J21Rik; Ab2-232; E4; KIAA0126; UBE4A; UBOX2; UFD2; UFD2b
UniProt ID: (Human) Q14139, (Rat) Q6P7A2, (Mouse) E9Q735
Entrez Gene ID: (Human) 9354, (Rat) 315608, (Mouse) 140630
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support