Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
May play a role as NF-kappa-B regulator. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ubiquitin domain-containing protein UBFD1; Ubiquitin-binding protein homolog
Gene Aliases: AI467302; D7Wsu105e; D7Wsu128e; UBFD1; UBPH
UniProt ID: (Human) O14562, (Mouse) Q78JW9
Entrez Gene ID: (Human) 56061, (Mouse) 28018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support