Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Bovine
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMSC-UbP; Bone marrow stromal cell ubiquitin-like protein; MGC14421; ubiquitin-like 7 (bone marrow stromal cell-derived); Ubiquitin-like protein 7; Ubiquitin-like protein SB132
Gene Aliases: 2300004C15Rik; BMSC-UbP; BMSCUBP; SB132; TCBA1; UBL7
UniProt ID: (Human) Q96S82, (Mouse) Q91W67
Entrez Gene ID: (Human) 84993, (Mouse) 69459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support