Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PPAATNSSKK MKFQGEPKKP PMNGYQKFSQ ELLSNGELNH LPLKERMVEI GSRWQRISQS QKEHYKKLAE EQQKQYKVHL DLWVKSLSPQ DRAAYKEYIS
UBF-1 encodes a member of the HMG-box DNA-binding protein family. The encoded protein plays a critical role in ribosomal RNA transcription as a key component of the pre-initiation complex, mediating the recruitment of RNA polymerase I to rDNA promoter regions. The encoded protein may also play important roles in chromatin remodeling and pre-rRNA processing, and its activity is regulated by both phosphorylation and acetylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3, 11 and X and the long arm of chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90-kDa nucleolus organizer region autoantigen; Autoantigen NOR-90; NOR90; Nucleolar transcription factor 1; transcription factor UBF; UBF-1; Upstream-binding factor 1
Gene Aliases: NOR-90; Tcfubf; UBF; UBF-1; UBF1; UBF2; UBTF
UniProt ID: (Human) P17480, (Rat) P25977
Entrez Gene ID: (Human) 7343, (Rat) 25574
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support