Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASAGGEDC ESPAPEADRP HQRPFLIGVS GGTASGKSTV CEKIMELLGQ NEVEQRQRKV VILSQDRFYK VLTAEQKAKA LKGQYNFDHP DAFDNDLMHR TLKNIVEGKT VEVPTYDFVT HSRLPETTVV YPADVVLFEG ILVFYSQEIR DMFHLRLFVD TDSDVRLSRR DKEVCRCDHP TRSGQYGCHQ PDRAAHPGHS EW (1-201 aa encoded by BC015547)
Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4- thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)- benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5- methylcytidine, and N(4)-anisoylcytidine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytidine monophosphokinase 1; UCK 1; uridine monophosphate kinase; Uridine monophosphokinase 1; Uridine-cytidine kinase 1
Gene Aliases: UCK1; Umpk; URK1
UniProt ID: (Human) Q9HA47, (Mouse) P52623
Entrez Gene ID: (Human) 83549, (Rat) 311864, (Mouse) 22245
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support