Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TSKRTIYTA GRPPWYNEHG TQSKEAFAIG LGGGSASGKT TVARMIIEAL DVPWVVLLSM DSFYKVLTEQ QQEQAAHNNF NFDHPDAFDF DLIISTLKKL KQGKSVKVPI YDFTTHSRKK DWKTLYGANV IIFEGIMAFA DKTLLELLDM KIFVDTDSDI RLVRRLRRDI SERGRDIEGV IKQYNKFVKP SFDQYIQPTM RLADIVVPRG SGNTVAIDLI VQHVHSQLEE GCAGLGTPVP PAAPDAERPE EHAAGTGHAH HHQVRAHLGT GRPRARAPAQ LLAPQGQGDQ SRRVHLLLQE TDAAAHRARA LLPALSGLRR TDPAGAGLCG QVLCGEADHR CVHSARR (74-419 aa encoded by BC033078)
Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UCK1-LIKE; uridine kinase-like 1; uridine-cytidine kinase 1-like 1; Uridine-cytidine kinase-like 1
Gene Aliases: 1110007H10Rik; F538; RGD1565465; UCK1L; UCKL1; URKL1
UniProt ID: (Human) Q9NWZ5, (Mouse) Q91YL3
Entrez Gene ID: (Human) 54963, (Mouse) 68556, (Rat) 499956
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support