Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detect a band around 28-33 kDa corresponding to UCP2 protein in mouse liver, skeletal muscle and pancreas. Some upper and lower bands also were observed due to the dimer formation and degradation of the UCP2 protein. This antibody was raised against the full-length UCP2 protein and may cross-react with UCP3. It is notable that UCP2 is known to have a very short half-life less than 1 hour. It is strongly recommended that fresh-made lysate be used for western blot assays.
Immunogen sequence: MVGFKATDV PPTATVKFLG AGTAACIADL ITFPLDTAKV RLQIQGESQG PVRATASAQY RGVMGTILTM VRTEGPRSLY NGLVAGLQRQ MSFASVRIGL YDSVKQFYTK GSEHASIGSR LLAGSTTGAL AVAVAQPTDV VKVRFQAQAR AGGGRRYQST VNAYKTIARE EGFRGLWKGT SPNVARNAIV NCAELVTYDL IKDALLKANL MTDDLPCHFT SAFGAGFCTT VIASPVDVVK TRYMNSALGQ YSSAGHCALT MLQKEGPRAF YKGFMPSFLR LGSWNVVMFV TYEQLKRALM AACTSREAPF (1-309 aa encoded by BC011737)
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dicarboxylate carrier SLC25A8; Mitochondrial uncoupling protein 2; Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein; uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH
UniProt ID: (Human) P55851
Entrez Gene ID: (Human) 7351
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support