Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TEEVKQDKD DIINIFSVAS GHLYERFLRI MMLSVLKNTK TPVKFWFLKN YLSPTFKEFI PYMANEYNFQ YELVQYKWPR WLHQQTEKQR IIWGYKILFL DVLFPLVVDK FLFVDADQIV RTDLKELRDF NLDGAPYGYT PFCDSRREMD GYRFWKSGYW ASHLAGRKYH ISALYVVDLK KFRKIAAGDR LRGQYQGLSQ DPNSLSNLDQ DLPNNMIHQV PIKSLPQEWL WCETWCDDAS KKRAKTIDLC NNPMTKEPKL EAAVRIVPEW QDYDQEIKQL QIRFQKEKET GALYKEKTKE PSREGPQKRE EL (1221-1531 aa encoded by B C041098)
UDP-glucose:glycoprotein glucosyltransferase (UGGT) is a 170 kDa lumenal endoplasmic reticulum (ER) resident protein that catalyzes monoglucosylation of high mannose oligosaccharides. UGGT catalyzes the transfer of glucose from UDP-glucose to Man7-9GlcNAc2-moiety of unfolded glycoproteins in a Ca2+ dependent reaction. UGGT binds unfolded glycoprotein via two elements on its substrate: a N-acetylglucosamine unit on the oligosaccharide and hydrophobic amino acids that are exposed in the denatured conformation. It is postulated that UGGT serves as a sensor that is able to distinguish between folded and denatured protein in the ER.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RUGT; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGT1
Gene Aliases: 0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; GT; HUGT1; rUGT1; UGCGL1; UGGT; UGGT1; UGT1; UGTR
UniProt ID: (Human) Q9NYU2, (Rat) Q9JLA3, (Mouse) Q6P5E4
Entrez Gene ID: (Human) 56886, (Rat) 171129, (Mouse) 320011
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support