Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPAKATNVV RLLLGSTALW LSQLGSGTVA ASKSVTAHLA AKWPETPLLL EASEFMAEES NEKFWQFLET VQELAIYKQT ESDYSYYNLI LKKAGQFLDN LHINLLKFAF SIRAYSPAIQ MFQQIAADEP PPDGCNAFVV IHKKHTCKIN EIKKLLKKAA SRTRPYLFKG DHKFPTNKEN LPVVILYAEM GTRTFSAFHK VLSEKAQNEE ILYVLRHYIQ KPSSRKMYLS GYGVELAIKS TEYKALDDTQ VKTVTNTTVE DETETNEVQG FLFGKLKS
UDP-glucose:glycoprotein glucosyltransferase (UGT) is a soluble protein of the endoplasmic reticulum (ER) that selectively reglucosylates unfolded glycoproteins, thus providing quality control for protein transport out of the ER.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP--Glc:glycoprotein glucosyltransferase 2; UDP-Glc:glycoprotein glucosyltransferase 2; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose ceramide glucosyltransferase-like 2; UDP-glucose:glycoprotein glucosyltransferase 2; UGT2
Gene Aliases: HUGT2; UGCGL2; UGGT2; UGT2
UniProt ID: (Human) Q9NYU1
Entrez Gene ID: (Human) 55757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support