Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: A-549, HT-29, MCF7, HeLa, Mouse liver. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome, Single-pass membrane protein.
Immunogen sequence: GKLLVVPMDG SHWFTMRSVV EKLILRGHEV VVVMPEVSWQ LGRSLNCTVK TYSTSYTLED LDREFKAFAH AQWKAQVRSI YSLLMGSYND IFDLFFSNCR SLFKDKKLVE YLKESSFDAV FLDPFDNCGL IVAKYFSLPS VVFARGILCH YLEEGAQCPA PLSYVPRILL GFSDAMTFKE RVRNHIMHLE EHLLCHRFFK NALEIASEIL QTPVTEYDLY SHTSI
UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. UGT1A9 is active on phenols.This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme encoded by this gene is active on phenols. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lugP4; UDP glucuronosyltransferase 1 family, polypeptide A9; UDP glycosyltransferase 1 family polypeptide A10; UDP glycosyltransferase 1 family polypeptide A11; UDP glycosyltransferase 1 family polypeptide A12; UDP glycosyltransferase 1 family, polypeptide A10; UDP glycosyltransferase 1 family, polypeptide A9; UDP-glucuronosyltransferase 1 family polypeptide A9s; UDP-glucuronosyltransferase 1-7; UDP-glucuronosyltransferase 1-9; UDP-glucuronosyltransferase 1-I; UDP-glucuronosyltransferase 1A9; UDPGT; UDPGT 1-9; UGT-1I; UGT1A12; UGT1A9; UGTP4
Gene Aliases: GNT1; HLUGP4; LUGP4; mUGTBr/P; UDPGT; UDPGT 1-9; UGT-1I; UGT1; UGT1-09; UGT1-9; UGT1.9; Ugt1a10; Ugt1a11; Ugt1a12; UGT1A9; UGT1A9S; UGT1AI; UGT1I; UGTP4
UniProt ID: (Human) O60656, (Mouse) Q62452
Entrez Gene ID: (Human) 54600, (Mouse) 394434, (Rat) 396552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support