Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SALHERGHHT VFLLSEGRDI APSNHYSLQR YPGIFNSTTS DAFLQSKMRN IFSGRLTAIE LFDILDHYTK NCDLMVGNHA LIQGLKKEKF DLLLVDPNDM CGFVIAHLLG VKYAVFSTGL WYPAEVGAPA
Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; Ceramide UDP-galactosyltransferase; Cerebroside synthase; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; uridine diphosphate glycosyltransferase 8
Gene Aliases: CGT; UGT4; UGT8
UniProt ID: (Human) Q16880
Entrez Gene ID: (Human) 7368
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support