Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ac2-121; E3 ubiquitin-protein ligase UHRF1; FLJ21925; Hnp95 Huhrf1; MGC138707; mUhrf1; Nuclear protein 95; Nuclear zinc finger protein Np95; RING-type E3 ubiquitin transferase UHRF1; ub; Ubiquitin-like PHD and RING finger domain-containing protein 1; Ubiquitin-like-containing PHD and RING finger domains protein 1
Gene Aliases: AL022808; ICBP90; Np95; RNF106; Uhrf1
UniProt ID: (Mouse) Q8VDF2
Entrez Gene ID: (Mouse) 18140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support