Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AGRADPHSL CYDITVIPKF RPGPRWCAVQ GQVDEKTFLH YDCGNKTVTP VSPLGKKLNV TTAWKAQNPV LREVVDILTE QLRDIQLENY TPKEPLTLQA RMSCEQKAEG HSSGSWQFSF DGQIFLLFDS EKRMWTTVHP GARKMKEKWE NDKVVAMSFH YFSMGDCIGW LEDFLMGMDS TLEPSAGAPL AMSSGTTQLR A (25-224 aa encoded by BC034689)
ULBP2 encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells. The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety, or secreted. Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells. This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALCAN-alpha; N2DL-2; NKG2D ligand 2; Retinoic acid early transcript 1H; UL16-binding protein 2
Gene Aliases: ALCAN-alpha; N2DL2; NKG2DL2; RAET1H; ULBP2; UNQ463/PRO791
UniProt ID: (Human) Q9BZM5
Entrez Gene ID: (Human) 80328
Molecular Function:
major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support