Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGPGWGPPR LDGFILTERL GSGTYATVYK AYAKKDTREV VAIKCVAKKS LNKASVENLL TEIEILKGIR HPHIVQLKDF QWDSDNIYLI MEFCAGGDLS RFIHTRRILP EKVARVFMQQ LASALQFLHE RNISHLDLKP QNILLSSLEK PHLKLADFGF AQHMSPWDEK HVLRGSPLYM APEMVCQRQY DARVDLWSMG VILYGETSFP CFSP
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; Unc-51-like kinase 3
Gene Aliases: ULK3
UniProt ID: (Human) Q6PHR2
Entrez Gene ID: (Human) 25989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support