Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGQPSLTWML MVVVASWFIT TAATDTSEAR WCSECHSNAT CTEDEAVTTC TCQEGFTGDG LTCVDLDECA IPGAHNCSAN SSCVNTPGSF SCVCPEGFRL SPGLGCTDVD ECAEPGLSHC HALATCVNVV GSYLCVCPAG YRGDGWHCEC SPGSCGPGLD CVPEGDALVC ADPCQAHRTL DEYWRSTEYG EGYACDTDLR GWYRPHPSSD EGIVSRKACA HWSGHCCLWD ASVQVKACAG GYYVYNLTAP PECHLAYCTD PSSVEGTCEE CSIDEDCKSN NGRWHCQCKQ DFNITDISLL EHRLECGAND MKVSLGKCQL KSLGFDKVFM YLSDSRCSGF NDRDNRDWVS VVTPARDGPC GTVLTRNETH ATYSNTLYLA DEIIIRDLNI KINFACSYPL DMKVSLKTAL QPMVSALNIR VGGTGMFTVR MALFQTPSYT QPYQGSSVTL STEAFLYVGT MLDGGDLSRF ALLMTNCYAT PSSNATDPLK YFIIQDRCPH TRDSTIQVVE NGESSQGRFS VQMFRFAGNY DLVYLHCEVY LCDTMNEKCK PTCSGTRFRS GSVIDQSRVL NLGPITRKGV QATVSRAFSS LGLLKVWLPL LLSATLTLTF Q
This gene encodes uromodulin, the most abundant protein in normal urine. Its excretion in urine follows proteolytic cleavage of the ectodomain of its glycosyl phosphatidylinosital-anchored counterpart that is situated on the luminal cell surface of the loop of Henle. Uromodulin may act as a constitutive inhibitor of calcium crystallization in renal fluids. Excretion of uromodulin in urine may provide defense against urinary tract infections caused by uropathogenic bacteria. Defects in this gene are associated with the autosomal dominant renal disorders medullary cystic kidney disease-2 (MCKD2) and familial juvenile hyperuricemic nephropathy (FJHN). These disorders are characterized by juvenile onset of hyperuricemia, gout, and progressive renal failure. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tamm-Horsfall urinary glycoprotein; THP; Uromodulin; uromucoid; uromucoid uromucoid
Gene Aliases: ADMCKD2; FJHN; HNFJ; HNFJ1; MCKD2; THGP; THP; UMOD
UniProt ID: (Human) P07911
Entrez Gene ID: (Human) 7369
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support