Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGVFCLGPW GLGRKLRTPG KGPLQLLSRL CGDHLQAIPA KKAPAGQEEP GTPPSSPLSA EQLDRIQRNK AAALLRLAAR NVPVGFGESW KKHLSGEFGK PYFIKLMGFV AEERKHYTVY PPPHQVFTWT QMCDIKDVKV VILGQDPYHG PNQAHGLCFS VQRPVPPPPS LENIYKELST DIEDFVHPGH GDLSGWAKQG VLLLNAVLTV RAHQANSHKE RGWEQFTDAV VSWLNQNSNG LVFLLWGSYA QKKGSAIDRK RHHVLQTAHP SPLSVYRGFF GCRHFSKTNE LLQKSGKKPI DWKEL (1-304 aa encoded by BC015205)
The human UNG gene encodes both nuclear (UNG2) and mitochondrial (UNG1) forms of uracil-DNA glycosylase. These forms are generated by alternative splicing and the use of two differentially regulated promoters, PA and PB. The cDNAs for UNG1 and UNG2 are of similar size (2061 and 2058 bp, respectively) and as a result the corresponding mRNAs are not resolved as two species in gel electrophoresis. Nuclear UNG2 differs from mitochondrial UNG1 in 44 amino acids of the N-terminal sequence that is not necessary for catalytic activity. A major role of the UNG gene products is to repair mutagenic U:G mispairs caused by cytosine deamination. For example, UNG2 removes misincorporated dUMP residues. The level and expression pattern of UNG1 and UNG2 differs between cell and tissue type. Additionally, the expression of UNG is cell cycle regulated. The expression and activity of UNG has been found in general to be higher in proliferating as compared to nonproliferating tissues and cells. UNG1 is a 304 amino acid protein. UNG2 is a 313 amino acid protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781L1143; UDG; UNG; Uracil-DNA glycosylase; uracil-DNA glycosylase 1, uracil-DNA glycosylase 2; uracil-DNA glycosylase nuclear isoform
Gene Aliases: DGU; HIGM4; HIGM5; UDG; UNG; UNG1; UNG15; UNG2
UniProt ID: (Human) P13051, (Mouse) P97931
Entrez Gene ID: (Human) 7374, (Mouse) 22256, (Rat) 304577
Molecular Function:
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support