Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
UQCRB encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This component is involved in redox-linked proton pumping.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 7; Complex III subunit VII; Cytochrome b-c1 complex subunit 7; mitochondrial ubiquinone-binding protein; QP-C; Ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase, complex III subunit VI
Gene Aliases: MC3DN3; QCR7; QP-C; QPC; UQBC; UQBP; UQCR6; UQCRB; UQPC
UniProt ID: (Human) P14927
Entrez Gene ID: (Human) 7381
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support