Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive Control: A549, 293, mouse kidney tissue lysate, 293T, LOVO, SiHa, rat skeletal muscle tissue, human colon cancer tissue, human appendix tissue, human kidney tissue, mouse cerebellum tissue.
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 5; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit Rieske, mitochondrial; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RISP; Ubiquinol-cytochrome c reductase iron-sulfur subunit
Gene Aliases: RIP1; RIS1; RISP; UQCR5; UQCRFS1
UniProt ID: (Human) P47985
Entrez Gene ID: (Human) 7386
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support