Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAPLSVEV EFGGGAELLF DGIKKHRVTL PGQEEPWDIR NLLIWIKKNL LKERPELFIQ GDSVRPGILV LINDADWELL GELDYQLQDQ DSVLFISTLH GG (1-101 aa encoded by BC003581)
Following the discovery of protein modification by the small, highly conserved ubiquitin polypeptide, a number of distinct ubiquitin-like proteins (Ubls) have been found to function as protein modifiers as well. These Ubls, which include SUMO, ISG15, Nedd8, and Atg8, function as critical regulators of many cellular processes, including transcription, DNA repair, signal transduction, autophagy, and cell-cycle control. A growing body of data also implicates the dysregulation of Ubl-substrate modification and mutations in the Ubl-conjugation machinery in the etiology and progression of a number of human diseases. URM1 is a ubiquitin-like modifier protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC2668; RP11-339B21.4; ubiquitin related modifier 1 homolog; Ubiquitin-related modifier 1; ubiquitin-related modifier 1 homolog; URM1
Gene Aliases: 2900073H19Rik; C9orf74; RGD1306599; URM1
UniProt ID: (Human) Q9BTM9, (Mouse) Q9D2P4
Entrez Gene ID: (Human) 81605, (Mouse) 68205, (Rat) 311840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support