Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
MDS032 belongs to the USE1 family and is a component of a SNARE complex which may be involved in targeting and fusion of Golgi-derived retrograde transport vesicles with the ER.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: embryonic development factor 2; p31; Protein D12; protein p31; putative MAPK activating protein PM26; Putative MAPK-activating protein PM26; Q-SNARE; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1 homolog; USE1-like protein; Vesicle transport protein USE1
Gene Aliases: 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; MDS032; P31; Q-snare; SLT1; USE1; USE1L
UniProt ID: (Human) Q9NZ43, (Mouse) Q9CQ56
Entrez Gene ID: (Human) 55850, (Mouse) 67023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support