Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAEGGAADLD TQRSDIATLL KTSLRKGDTW YLVDSRWFKQ WKKYVGFDSW DKYQMGDQNV YPGPIDNSGL LKDGDAQSLK EHLIDELDYI LLPTEGWNKL VSWYTLMEGQ EPIARKVVEQ GMFVKHCKVE VYLTELKLCE NGNMNNVVTR RFSKADTIDT IEKEIRKIFS IPDEKETRLW NKYMSNTFEP LNKPDSTIQD AGLYQGQVLV IEQKNEDGTW PRGPSTPKKP LEQSC
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 15; MGC131982; MGC149838; MGC74854; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; Unph4
Gene Aliases: KIAA0529; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y5B5
Entrez Gene ID: (Human) 9958
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support