Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: AGGSSVKSVC KNTGDDEAER GHQPPASQQP NANTTGKEQL VTKDPASAKH SLLSARKSKS SQLDSGVPSS PGGRQSAEKS SKKLSSSMQT SARPSQKPQ
May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 31; Ubiquitin carboxyl-terminal hydrolase 31; ubiquitin specific proteinase 31; Ubiquitin thioesterase 31; ubiquitin thiolesterase 31; Ubiquitin-specific-processing protease 31
Gene Aliases: KIAA1203; USP31
UniProt ID: (Human) Q70CQ4
Entrez Gene ID: (Human) 57478
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support