Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDQCYFENSL STECIIRSAS RSDGCQMPKL FCQNLPPPLP PKKYAITSVP QSEKSESTPD VKLTEVFKAT SHLPKHSLST ASEPSLEVST HMNDERHKET FQVRECFGNT PNCPSSSSTN DFQANSGAID AFCQPELDSI STCPNETVSL TTYFSVDSCM TDTYRLKYHQ RPKLSFPESS GFCNNSLS
Tight junction-associated protein that is involved in the survival of auditory hair cells and hearing. Maybe by modulating the barrier properties and mechanical stability of tight junctions. Has no peptidase activity (PubMed:14715245). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive ubiquitin carboxyl-terminal hydrolase 53; Inactive ubiquitin-specific peptidase 53; ubiquitin specific protease 53; ubiquitin specific proteinase 53; USP53
Gene Aliases: KIAA1350; USP53
UniProt ID: (Human) Q70EK8
Entrez Gene ID: (Human) 54532
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support