Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MTATTRGSPV GGNDNQGQAP DGQSQPPLQQ NQTSSPDSSN ENSPATPPDE QGQGDAPPQL EDEEPAFPHT DLAKLDDMIN RPRWVVPVLP
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme FAF-X; Drosophila fat facets related, X-linked; Fat facets in mammals; fat facets protein related, X-linked; Fat facets protein-related, X-linked; hFAM; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); Ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; Ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X
Gene Aliases: DFFRX; FAF; FAM; MRX99; MRXS99F; USP9; USP9X
UniProt ID: (Human) Q93008
Entrez Gene ID: (Human) 8239
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support