Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, H1299, HeLaS3.
Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (82%), Rhesus Monkey (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Uronyl 2-sulfotransferase transfers sulfate to the 2-position of uronyl residues, such as iduronyl residues in dermatan sulfate and glucuronyl residues in chondroitin sulfate (Kobayashi et al., 1999 [PubMed 10187838]).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dermatan/chondroitin sulfate 2-sulfotransferase; Uronyl 2-sulfotransferase; uronyl-2-sulfotransferase
Gene Aliases: 2OST; DS2ST; UST
UniProt ID: (Human) Q9Y2C2
Entrez Gene ID: (Human) 10090
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support