Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Involved in nucleolar processing of pre-18S ribosomal RNA. Target protein may have a role in maintainance of neuronal function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable U3 small nucleolar RNA-associated protein 11; U3 snoRNA-associated protein 11; UTP11-like protein; UTP11-like, U3 small nucleolar ribonucleoprotein
Gene Aliases: CGI-94; CGI94; HDCMB12P; UTP11; UTP11L
UniProt ID: (Human) Q9Y3A2
Entrez Gene ID: (Human) 51118
Molecular Function: RNA metabolism protein nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support