Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HT-1080, Mouse kidney, Mouse brain, Rat kidney, Rat brain. The target is usually found in the following locations: Secreted.
Immunogen sequence: GSNELHQVPS NCDCLNGGTC VSNKYFSNIH WCNCPKKFGG QHCEIDKSKT CYEGNGHFYR GKASTDTMGR PCLPWNSATV LQQTYHAHRS DALQLGLGKH NYCRNPDNRR RPWCYVQVGL KPLVQECMVH DCADGKKPSS PPEELKFQCG QKTLRPRFKI I
This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antagonist of uPA; HGNC:9052; plasminogen activator; plasminogen activator, urinary; U-plasminogen activator; uPA; UPAR; Urinary plasminogen activator, urokinase; URKR; Urokinase-type plasminogen activator
Gene Aliases: ATF; BDPLT5; PLAU; QPD; u-PA; UPA; UPAM; URK
UniProt ID: (Human) P00749, (Mouse) P06869
Entrez Gene ID: (Human) 5328, (Rat) 25619, (Mouse) 18792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support