Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KMFRHTDSL FPILLQTLSD ESDEVILKDL EVLAEIASSP AGQTDDPGPL DGPDLQASHS ELQVPTPGRA GLLNTSGTKG LECSPSTPTM NSYFYKFMIN LLKRFSSERK LLEVRGPFII RQLCLLLNAE NIFHSMADIL LREEDLKFAS TMVHALNTIL LTSTELFQLR NQLKDLKTLE SQNLFCCLYR SWCHNPVTTV SLCFLTQNYR HAYDLIQKFG DLEVTVDFLA EVDKLVQLIE CPIFTYLRLQ LLDVKNNPYL IKALYGLLML LPQSSAFQLL SHRLQCVPNP ELLQTEDSLK AAPKSQKADS PSIDYAELLQ HFEKVQNKHL EVRHQRSGRG DHLDRRVVL (228-576 aa encoded by BC007214)
Vac14 a putative cytoskeletal interacting protein; mutation of gene may be associated with lethal communicating hydrocephalus with perinatal onset.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein VAC14 homolog; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; Tax1-binding protein 2; Vac14 homolog; vacuole 14 protein; vacuole morphology 14
Gene Aliases: AA959718; ArPIKfyve; BC032215; D8Wsu151e; ingls; TAX1BP2; TRX; VAC14
UniProt ID: (Human) Q08AM6, (Rat) Q80W92, (Mouse) Q80WQ2
Entrez Gene ID: (Human) 55697, (Rat) 307842, (Mouse) 234729
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support